Coelenterazine ip, Sigma-Aldrich®
Lieferant: Merck
Achtung
Synonyme: Coelenterazine IP, CLZN-ip
M-Clarity™ Qualitätsstufe = MQ200
C3605-50UG
SIALC3605-50UGEA 157 EUR
SIALC3605-50UG
Coelenterazine ip, Sigma-Aldrich®
Coelenterazine ip
Almost 50 times higher luminescence intensity than native coelenteraztine (Prod. No. C2230) but with a slower response time.