3-Chlor-2,5-dimethylpyrazin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

C38400-25G
SIALC38400-25GEA 65.3 EUR
SIALC38400-25G
3-Chlor-2,5-dimethylpyrazin, Sigma-Aldrich®
3-Chlor-2,5-dimethylpyrazin
Formel: C₆H₇ClN₂
Molecular Weight: 142,5900 g/mol
MDL: MFCD00006143
CAS-Nummer: 95-89-6
EINECS: 202-463-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR