D-Cycloserin, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
D(+)-Cycloserin, D-4-Aminoisoxazolidin-3-on, (R)-(+)-Cycloserin
M-Clarity™ Qualitätsstufe = MQ200
C3909-100MG
C3909-250MG
SIALC3909-100MGEA
206
EUR
SIALC3909-100MG
SIALC3909-250MG
D-Cycloserin, Sigma-Aldrich®
D-Cycloserin
Formel:
C₃H₆N₂O₂ Molecular Weight: 102.09 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00005353 CAS-Nummer: 68-41-7 EINECS: 200-688-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...