Chrom(III)picolinat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Picolinic acid chromium salt, Chromium picolinate

C4124-5G
SIALC4124-5GEA 350 EUR
SIALC4124-5G
Chrom(III)picolinat, Sigma-Aldrich®
Chrom(III)picolinat
Formel: C₁₈H₁₂CrN₃O₆
MDL: MFCD00068715
CAS-Nummer: 14639-25-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR