4-(2-Chlorethyl)morpholin Hydrochlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: N-(2-Chloroethyl)morpholine hydrochloride, N-(2-Chloroethyl)-Morpholinium Chloride

M-Clarity™ Qualitätsstufe = MQ200

C42203-100G
SIALC42203-100GEA 39.4 EUR
SIALC42203-100G
4-(2-Chlorethyl)morpholin Hydrochlorid, Sigma-Aldrich®
4-(2-Chlorethyl)morpholin Hydrochlorid
Formel: C₆H₁₃Cl₂NO
Molecular Weight: 186.08 g/mol
MDL: MFCD00012797
CAS-Nummer: 3647-69-6
EINECS: 222-881-2
UN: 2923
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR