Creatinin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Amino-1-methyl-1,5-dihydro-4H-imidazol-4-one

MQ200

C4255-100G C4255-10G C4255-1KG C4255-25G C4255-10MG
SIALC4255-100GEA 163 EUR
SIALC4255-100G SIALC4255-10G SIALC4255-1KG SIALC4255-25G SIALC4255-10MG
Creatinin, Sigma-Aldrich®
Creatinin
Formel: C₄H₇N₃O
Molecular Weight: 113.12 g/mol
MDL: MFCD00059730
CAS-Nummer: 60-27-5
EINECS: 200-466-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR