5-Chlor-2-hydroxybenzophenon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (5-Chloro-2-hydroxyphenyl)(phenyl)methanone

C44702-5G
SIALC44702-5GEA 66.9 EUR
SIALC44702-5G
5-Chlor-2-hydroxybenzophenon, Sigma-Aldrich®
5-Chlor-2-hydroxybenzophenon
Formel: C₁₃H₉ClO₂
Molecular Weight: 232,6600 g/mol
MDL: MFCD00020134
CAS-Nummer: 85-19-8
EINECS: 201-592-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR