4-Chlor-4'-hydroxybutyrophenon, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

C44907-5G
SIALC44907-5GEA 86.9 EUR
SIALC44907-5G
4-Chlor-4'-hydroxybutyrophenon, Sigma-Aldrich®
4-Chlor-4'-hydroxybutyrophenon
Formel: C₁₀H₁₁ClO₂
MDL: MFCD00002362
CAS-Nummer: 7150-55-2
EINECS: 230-485-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR