5-Chlor-2-indolcarbonsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Chloro-1H-indole-2-carboxylic acid

M-Clarity™ Qualitätsstufe = MQ100

C47809-1G
SIALC47809-1GEA 43.1 EUR
SIALC47809-1G
5-Chlor-2-indolcarbonsäure, Sigma-Aldrich®
5-Chloroindole-2-carboxylic acid
Formel: C₉H₆ClNO₂
Molecular Weight: 195,6 g/mol
Schmelzpunkt: 291…292 °C
MDL: MFCD00005613
CAS-Nummer: 10517-21-2
EINECS: 234-050-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR