2-Chlor-6-methoxy-3-nitropyridin, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

C49909-10G
SIALC49909-10GEA 70.6 EUR
SIALC49909-10G
2-Chlor-6-methoxy-3-nitropyridin, Sigma-Aldrich®
2-Chloro-6-methoxy-3-nitropyridine
Formel: C₆H₅ClN₂O₃
Molecular Weight: 188,57 g/mol
MDL: MFCD00130268
CAS-Nummer: 38533-61-8
EINECS: 253-989-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR