4-(4-Chlorphenyl)-4-piperidinol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

C66056-5G
SIALC66056-5GEA 87.4 EUR
SIALC66056-5G
4-(4-Chlorphenyl)-4-piperidinol, Sigma-Aldrich®
4-(4-Chlorphenyl)-4-piperidinol
Formel: C₁₁H₁₄ClNO
MDL: MFCD00006001
CAS-Nummer: 39512-49-7
EINECS: 254-479-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR