Cytochalasin A (aus Dreschslera dematioidea), Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
C6637-1MG
SIALC6637-1MGEA
136
EUR
SIALC6637-1MG
Cytochalasin A (aus Dreschslera dematioidea), Sigma-Aldrich®
Cytochalasin A
Formel:
C₂₉H₃₅NO₅ Molecular Weight: 477,6 g/mol |
MDL:
MFCD00005935 CAS-Nummer: 14110-64-6 EINECS: 237-964-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...