Cytochalasin B (aus Dreschslera dematioidea), Sigma-Aldrich®
Lieferant: Merck
Synonyme:
7(S),20(R)-Dihydroxy-16(R)-methyl-10-phenyl-24-oxa[14]cytochalasa-6(12),13(E),21(E)-triene-1,23-dione
M-Clarity™ Qualitätsstufe = MQ300
C6762-1MG
C6762-25MG
C6762-50MG
C6762-5MG
C6762-10MG
SIALC6762-1MGEA
105
EUR
SIALC6762-1MG
SIALC6762-25MG
SIALC6762-50MG
SIALC6762-5MG
SIALC6762-10MG
Cytochalasin B (aus Dreschslera dematioidea), Sigma-Aldrich®
Cytochalasin B
Formel:
C₂₉H₃₇NO₅ Molecular Weight: 479.62 g/mol |
MDL:
MFCD00077704 CAS-Nummer: 14930-96-2 EINECS: 239-000-2 UN: 1544 ADR: 6.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...