L(+)-Cystein Hydrochlorid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

C6852-100G C6852-1KG C6852-25G
SIALC6852-100GEA 143 EUR
SIALC6852-100G SIALC6852-1KG SIALC6852-25G
L(+)-Cystein Hydrochlorid, Sigma-Aldrich®
L(+)-Cystein Hydrochlorid
Formel: HSCH₂CH(NH₂)COOH·HCl
Schmelzpunkt: 175 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00064553
CAS-Nummer: 52-89-1
EINECS: 200-157-7
Merck Index: 13,02810

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR