5-Chlor-2'-deoxyuridin, Sigma-Aldrich®

Lieferant: Merck

C6891-100MG
SIALC6891-100MGEA 262 EUR
SIALC6891-100MG
5-Chlor-2'-deoxyuridin, Sigma-Aldrich®
5-Chlor-2'-deoxyuridin
Formel: C₉H₁₁ClN₂O₅
MDL: MFCD00006531
CAS-Nummer: 50-90-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR