N-Cbz-Glycin (Z-Gly-OH), Sigma-Aldrich®

Lieferant: Merck

Synonyme: N-Carbobenzyloxyglycine, N-Benzyloxycarbonylglycine, Z-Gly-OH, 2-(((Benzyloxy)carbonyl)amino)acetic acid

M-Clarity™ Qualitätsstufe = MQ200

C7206-100G C7206-25G C7206-500G
SIALC7206-100GEA 156 EUR
SIALC7206-100G SIALC7206-25G SIALC7206-500G
N-Cbz-Glycin (Z-Gly-OH), Sigma-Aldrich®
N-Cbz-Glycin (Z-Gly-OH)
Formel: C₁₀H₁₁NO₄
Molecular Weight: 209.2 g/mol
MDL: MFCD00002691
CAS-Nummer: 1138-80-3
EINECS: 214-516-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR