N-Cbz-Glycin (Z-Gly-OH), Sigma-Aldrich®
Lieferant: Merck
Synonyme:
N-Carbobenzyloxyglycine, N-Benzyloxycarbonylglycine, Z-Gly-OH, 2-(((Benzyloxy)carbonyl)amino)acetic acid
M-Clarity™ Qualitätsstufe = MQ200
C7206-100G
C7206-25G
C7206-500G
SIALC7206-100GEA
156
EUR
SIALC7206-100G
SIALC7206-25G
SIALC7206-500G
N-Cbz-Glycin (Z-Gly-OH), Sigma-Aldrich®
N-Cbz-Glycin (Z-Gly-OH)
Formel:
C₁₀H₁₁NO₄ Molecular Weight: 209.2 g/mol |
MDL:
MFCD00002691 CAS-Nummer: 1138-80-3 EINECS: 214-516-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...