5-Cholesten-3-on, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

C75004-500MG
SIALC75004-500MGEA 280 EUR
SIALC75004-500MG
5-Cholesten-3-on, Sigma-Aldrich®
5-Cholesten-3-one
Formel: C₂₇H₄₄O
Molecular Weight: 384.65 g/mol
CAS-Nummer: 601-54-7
EINECS: 210-004-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR