Cholinchlorid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(2-Hydroxyethyl)trimethylammoniumchlorid
MQ200
C7527-500G
C7527-100G
C7527-1KG
SIALC7527-500GEA
164
EUR
SIALC7527-500G
SIALC7527-100G
SIALC7527-1KG
Cholinchlorid, Sigma-Aldrich®
Cholinchlorid
Formel:
HOH₂CCH₂N(CH₃)₃Cl Molecular Weight: 139.62 g/mol Schmelzpunkt: 244…247 °C Dichte: ∼1.205 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011721 CAS-Nummer: 67-48-1 EINECS: 200-655-4 Merck Index: 13,02226 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...