Cycloheximid, Sigma-Aldrich®
Lieferant: Merck
|
Gefahr
|
Synonyme:
Naramycin A, CHX, 4-[(2R)-2-[(1S,3S,5S)-3,5-Dimethyl-2-oxocyclohexyl]-2-hydroxyethyl]piperidin-2,6-dion, Actidion
M-Clarity™ Qualitätsstufe = MQ200
C7698-1G
C7698-5G
SIALC7698-1GEA
103
EUR
SIALC7698-1G
SIALC7698-5G
Cycloheximid, Sigma-Aldrich®
Cycloheximid
Formel:
C₁₅H₂₃NO₄ Molecular Weight: 281.35 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00082346 CAS-Nummer: 66-81-9 EINECS: 200-636-0 UN: 2811 ADR: 6.1 I |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...