Cumarin-3-carbonsäure, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2-Oxo-2H-chromene-3-carboxylic acid
M-Clarity™ Qualitätsstufe = MQ100
C85603-25G
SIALC85603-25GEA
165
EUR
SIALC85603-25G
Cumarin-3-carbonsäure, Sigma-Aldrich®
Cumarin-3-carbonsäure
Formel:
C₁₀H₆O₄ Molecular Weight: 190,16 g/mol |
MDL:
MFCD00006852 CAS-Nummer: 531-81-7 EINECS: 208-518-0 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...