4-(2-Phenyl-2-propanyl)phenol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-(2-Phenylpropan-2-yl)phenol

M-Clarity™ Qualitätsstufe = MQ100

C87800-100G
SIALC87800-100GEA 56.6 EUR
SIALC87800-100G
4-(2-Phenyl-2-propanyl)phenol, Sigma-Aldrich®
4-Cumylphenol
Formel: C₁₅H₁₆O
Molecular Weight: 212,29 g/mol
MDL: MFCD00002365
CAS-Nummer: 599-64-4
EINECS: 209-968-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR