DL-Carnitin Hydrochlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Hydroxy-4-(trimethylammonio)butanoate hydrochloride

M-Clarity™ Qualitätsstufe = MQ200

C9500-100G
SIALC9500-100GEA 133 EUR
SIALC9500-100G
DL-Carnitin Hydrochlorid, Sigma-Aldrich®
DL-Carnitin Hydrochlorid
Formel: C₇H₁₆ClNO₃
Molecular Weight: 197,66 g/mol
MDL: MFCD00011904
CAS-Nummer: 461-05-2
EINECS: 207-309-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR