6-Chloruracil, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 6-Chlor-2,4-dihydroxypyrimidin, 4-Chlor-2,6-dihydroxypyrimidin, 4-Chloruracil, 6-Chlor-2,4(1H,3H)-pyrimidindion

C9525-10G
SIALC9525-10GEA 505 EUR
SIALC9525-10G
6-Chloruracil, Sigma-Aldrich®
6-Chloruracil
Formel: C₄H₃ClN₂O₂
Molecular Weight: 146,53 g/mol
MDL: MFCD00014595
CAS-Nummer: 4270-27-3
EINECS: 224-258-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR