Density Standards, ASTM D4052, at 15 °C, certified reference material, Supelco®
Lieferant: Merck
CRMUDEKR-250ML CRMDEGA-250ML CRMDEGO-250ML CRMDEKR-250ML CRMUDEGA-250ML CRMUDEGO-250ML
SIALCRMUDEKR-250MLEA 334 EUR
SIALCRMUDEKR-250ML SIALCRMDEGA-250ML SIALCRMDEGO-250ML SIALCRMDEKR-250ML SIALCRMUDEGA-250ML SIALCRMUDEGO-250ML
Density Standards, ASTM D4052, at 15 °C, certified reference material, Supelco®
Standards Dichtestandards
Bitte beachten Limited shelf life, expiry date on the label.