FIA aromatics standards, ASTM D 1319, Supelco®
Lieferant: Merck
CRMFIKR-250ML
SIALCRMFIKR-250MLEA
338
EUR
SIALCRMFIKR-250ML
FIA aromatics standards, ASTM D 1319, Supelco®
Standards
Mineralölindustrie-Standards
Fuels Testing Certified Reference Material for the calibration and verification of laboratory instruments used in testing petroleum and derivative products for hydrocarbon types in liquid petroleum products by fluorescent indicator adsorption ASTM D1319.