Natrium-1-decansulfonat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Sodium decane-1-sulphonate, Decane-1-sulphonic acid sodium salt, 1-Decanesulphonic acid, sodium salt

D0181-5G
SIALD0181-5GEA 288 EUR
SIALD0181-5G
Natrium-1-decansulfonat, Sigma-Aldrich®
Natrium-1-decansulfonat
Formel: C₁₀H₂₁NaO₃S
Molecular Weight: 244,33 g/mol
Schmelzpunkt: > 300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007526
CAS-Nummer: 13419-61-9
EINECS: 236-525-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR