1,3-Diethylharnstoff, Sigma-Aldrich®

Lieferant: Merck

Synonyme: N,N'-Diethylurea

M-Clarity™ Qualitätsstufe = MQ100

D101087-25G
SIALD101087-25GEA 80 EUR
SIALD101087-25G
1,3-Diethylharnstoff, Sigma-Aldrich®
1,3-Diethylharnstoff
Formel: C₅H₁₂N₂O
Molecular Weight: 116.16 g/mol
MDL: MFCD00009028
CAS-Nummer: 623-76-7
EINECS: 210-811-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR