1,3-Dihydroxyaceton dimer, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2,5-Dihydroxy-1,4-dioxane-2,5-dimethanol
M-Clarity™ Qualitätsstufe = MQ100
D107204-100G
D107204-500G
D107204-5G
SIALD107204-100GEA
70.4
EUR
SIALD107204-100G
SIALD107204-500G
SIALD107204-5G
1,3-Dihydroxyaceton dimer, Sigma-Aldrich®
1,3-Dihydroxyaceton dimer
Formel:
C₆H₁₂O₆ Molecular Weight: 180.16 g/mol |
MDL:
MFCD00051019 CAS-Nummer: 62147-49-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...