1,3-Dihydroxyaceton dimer, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,5-Dihydroxy-1,4-dioxane-2,5-dimethanol

M-Clarity™ Qualitätsstufe = MQ100

D107204-100G D107204-500G D107204-5G
SIALD107204-100GEA 70.4 EUR
SIALD107204-100G SIALD107204-500G SIALD107204-5G
1,3-Dihydroxyaceton dimer, Sigma-Aldrich®
1,3-Dihydroxyaceton dimer
Formel: C₆H₁₂O₆
Molecular Weight: 180.16 g/mol
MDL: MFCD00051019
CAS-Nummer: 62147-49-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR