2,2'-Dihydroxybenzophenon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Bis(2-hydroxyphenyl)methanone

M-Clarity™ Qualitätsstufe = MQ100

D110205-5G
SIALD110205-5GEA 124 EUR
SIALD110205-5G
2,2'-Dihydroxybenzophenon, Sigma-Aldrich®
2,2'-Dihydroxybenzophenon
Formel: C₁₃H₁₀O₃
Molecular Weight: 214,2167 g/mol
MDL: MFCD00002217
CAS-Nummer: 835-11-0
EINECS: 212-639-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR