5,7-Diiod-8-chinolinol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

D123609-100G
SIALD123609-100GEA 247 EUR
SIALD123609-100G
5,7-Diiod-8-chinolinol, Sigma-Aldrich®
5,7-Diiod-8-chinolinol
Formel: C₉H₅I₂NO
MDL: MFCD00006789
CAS-Nummer: 83-73-8
EINECS: 201-497-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR