3,3'-Diaminobenzidin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: DAB, Biphenyl-3,3',4,4'-tetrayltetraamin

MQ200

D12384-1G D12384-25G D12384-5G
SIALD12384-1GEA 30.8 EUR
SIALD12384-1G SIALD12384-25G SIALD12384-5G
3,3'-Diaminobenzidin, Sigma-Aldrich®
3,3'-Diaminobenzidin
Formel: C₁₂H₁₄N₄
Molecular Weight: 214.27 g/mol
MDL: MFCD00007725
CAS-Nummer: 91-95-2
EINECS: 202-110-6

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR