2',3'-Didesoxyadenosin, Sigma-Aldrich®

Lieferant: Merck

D1285-25MG D1285-5MG
SIALD1285-25MGEA 744 EUR
SIALD1285-25MG SIALD1285-5MG
2',3'-Didesoxyadenosin, Sigma-Aldrich®
2',3'-Didesoxyadenosin
Formel: C₁₀H₁₃N₅O₂
MDL: MFCD00010534
CAS-Nummer: 4097-22-7
EINECS: 223-853-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR