2,5-Dimercapto-1,3,4-thiadiazol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1,3,4-Thiadiazole-2,5-dithiol

M-Clarity™ Qualitätsstufe = MQ100

D129003-100G
SIALD129003-100GEA 92.1 EUR
SIALD129003-100G
2,5-Dimercapto-1,3,4-thiadiazol, Sigma-Aldrich®
2,5-Dimercapto-1,3,4-thiadiazole
Formel: C₂H₂N₂S₃
Molecular Weight: 150,25 g/mol
MDL: MFCD00003103
CAS-Nummer: 1072-71-5
EINECS: 214-014-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR