2',4'-Dimethoxyacetophenon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-(2,4-Dimethoxyphenyl)ethan-1-on

M-Clarity™ Qualitätsstufe = MQ100

D129402-25G
SIALD129402-25GEA 37 EUR
SIALD129402-25G
2',4'-Dimethoxyacetophenon, Sigma-Aldrich®
2',4'-Dimethoxyacetophenon
Formel: C₁₀H₁₂O₃
Molecular Weight: 180.2 g/mol
MDL: MFCD00008727
CAS-Nummer: 829-20-9
EINECS: 212-587-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR