(Dimethylamino)acetaldehyddiethylacetal, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,2-Diethoxy-N,N-dimethylethanamine

MQ100

D138800-25G
SIALD138800-25GEA 115 EUR
SIALD138800-25G
(Dimethylamino)acetaldehyddiethylacetal, Sigma-Aldrich®
(Dimethylamino)acetaldehyddiethylacetal
Formel: C₈H₁₉NO₂
Molecular Weight: 161.25 g/mol
MDL: MFCD00009232
CAS-Nummer: 3616-56-6
EINECS: 222-800-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR