4-(Dimethylamino)benzonitril, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-(Dimethylamino) benzonitrile

M-Clarity™ Qualitätsstufe = MQ200

D139505-25G
SIALD139505-25GEA 141 EUR
SIALD139505-25G
4-(Dimethylamino)benzonitril, Sigma-Aldrich®
4-(Dimethylamino)benzonitril
Formel: C₉H₁₀N₂
Molecular Weight: 146,19 g/mol
MDL: MFCD00001815
CAS-Nummer: 1197-19-9
EINECS: 214-819-8
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR