(2-Chlorpropyl)dimethylammoniumchlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: N-(2-Chloropropyl)-N,N-dimethylammonium chloride, 2-Chloro-N,N-dimethylpropan-1-amine hydrochloride, 2-Dimethylaminoisopropyl chloride hydrochloride

M-Clarity™ Qualitätsstufe = MQ200

D142409-100G
SIALD142409-100GEA 31.4 EUR
SIALD142409-100G
(2-Chlorpropyl)dimethylammoniumchlorid, Sigma-Aldrich®
2-Chloropropyldimethylammonium chloride
Formel: C₅H₁₃Cl₂N
Molecular Weight: 158,07 g/mol
MDL: MFCD00012534
CAS-Nummer: 4584-49-0
EINECS: 224-971-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR