3-(Dimethylamino)propiophenon Hydrochlorid, Sigma-Aldrich®

Lieferant: Merck

D144800-25G
SIALD144800-25GEA 44.5 EUR
SIALD144800-25G
3-(Dimethylamino)propiophenon Hydrochlorid, Sigma-Aldrich®
3-(Dimethylamino)propiophenon Hydrochlorid
Formel: C₁₁H₁₆ClNO
Molecular Weight: 213,71 g/mol
MDL: MFCD00012481
CAS-Nummer: 879-72-1
EINECS: 212-909-1
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR