(3-Chlorpropyl)dimethylammoniumchlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Dimethylaminopropylchloride hydrochloride, 3-Dimethylaminopropyl chloride hydrochloride, N-(3-Chloropropyl)-N,N-dimethylammonium chloride, 3-Chloro-N,N-dimethylpropan-1-amine hydrochloride

M-Clarity™ Qualitätsstufe = MQ200

D145203-100G
SIALD145203-100GEA 50.8 EUR
SIALD145203-100G
(3-Chlorpropyl)dimethylammoniumchlorid, Sigma-Aldrich®
3-Chloropropyldimethylammonium chloride
Formel: C₅H₁₃Cl₂N
Molecular Weight: 158,07 g/mol
MDL: MFCD00012521
CAS-Nummer: 5407-04-5
EINECS: 226-467-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR