3,4-Dimethylbenzophenon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (3,4-Dimethylphenyl)(phenyl)methanone

M-Clarity™ Qualitätsstufe = MQ100

D149675-50G
SIALD149675-50GEA 161 EUR
SIALD149675-50G
3,4-Dimethylbenzophenon, Sigma-Aldrich®
3,4-Dimethylbenzophenon
Formel: C₁₅H₁₄O
Molecular Weight: 210,28 g/mol
MDL: MFCD00008525
CAS-Nummer: 2571-39-3
EINECS: 219-916-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR