2,6-Dimethyl-p-benzochinon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,6-Dimethylcyclohexa-2,5-diene-1,4-dione

M-Clarity™ Qualitätsstufe = MQ100

D149705-1G D149705-5G
SIALD149705-1GEA 63.4 EUR
SIALD149705-1G SIALD149705-5G
2,6-Dimethyl-p-benzochinon, Sigma-Aldrich®
2,6-Dimethyl-p-benzochinon
Formel: C₈H₈O₂
MDL: MFCD00001605
CAS-Nummer: 527-61-7
EINECS: 208-420-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR