N,N-Dimethylcarbamoylchlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Dimethylcarbamyl chloride

M-Clarity™ Qualitätsstufe = MQ200

D152803-100G D152803-500G D152803-5G
SIALD152803-100GEA 42.2 EUR
SIALD152803-100G SIALD152803-500G SIALD152803-5G
N,N-Dimethylcarbamoylchlorid, Sigma-Aldrich®
N,N-Dimethylcarbamoylchlorid
Formel: C₃H₆ClNO
MDL: MFCD00000635
CAS-Nummer: 79-44-7
EINECS: 201-208-6
UN: 2262
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR