2,2-Dimethyl-1,3-dioxolan, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

D155624-25G
SIALD155624-25GEA 102 EUR
SIALD155624-25G
2,2-Dimethyl-1,3-dioxolan, Sigma-Aldrich®
2,2-Dimethyl-1,3-dioxolan
Formel: C₅H₁₀O₂
Molecular Weight: 102,13 g/mol
MDL: MFCD00014106
CAS-Nummer: 2916-31-6
EINECS: 220-842-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR