3,3-Dimethylglutarsäureanhydrid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4,4-Dimethyldihydro-2H-pyran-2,6(3H)-dione

M-Clarity™ Qualitätsstufe = MQ200

D159808-100G
SIALD159808-100GEA 711 EUR
SIALD159808-100G
3,3-Dimethylglutarsäureanhydrid, Sigma-Aldrich®
3,3-Dimethylglutarsäureanhydrid
Formel: C₇H₁₀O₃
Molecular Weight: 142.15 g/mol
MDL: MFCD00006684
CAS-Nummer: 4160-82-1
EINECS: 223-998-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR