3,3-Dimethylglutarimid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4,4-dimethylpiperidine-2,6-dione

M-Clarity™ Qualitätsstufe = MQ100

D160008-25G
SIALD160008-25GEA 93.3 EUR
SIALD160008-25G
3,3-Dimethylglutarimid, Sigma-Aldrich®
3,3-Dimethylglutarimid
Formel: C₇H₁₁NO₂
Molecular Weight: 141,17 g/mol
MDL: MFCD00006671
CAS-Nummer: 1123-40-6
EINECS: 214-373-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR