N,N'-Dimethyl-1,6-diaminohexan, Sigma-Aldrich®

Lieferant: Merck

Synonyme: N1,N6-Dimethylhexane-1,6-diamine

M-Clarity™ Qualitätsstufe = MQ100

D161101-10G
SIALD161101-10GEA 257 EUR
SIALD161101-10G
N,N'-Dimethyl-1,6-diaminohexan, Sigma-Aldrich®
N,N'-Dimethyl-1,6-diaminohexan
Formel: C₈H₂₀N₂
Molecular Weight: 144,2578 g/mol
MDL: MFCD00008293
CAS-Nummer: 13093-04-4
EINECS: 236-005-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR