N,O-Dimethylhydroxylamin Hydrochlorid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

D163708-100G D163708-25G D163708-5G
SIALD163708-100GEA 272 EUR
SIALD163708-100G SIALD163708-25G SIALD163708-5G
N,O-Dimethylhydroxylamin Hydrochlorid, Sigma-Aldrich®
N,O-Dimethylhydroxylamin Hydrochlorid
Formel: C₂H₈ClNO
MDL: MFCD00012485
CAS-Nummer: 6638-79-5
EINECS: 229-642-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR