5'-Desoxyadenosin, Sigma-Aldrich®

Lieferant: Merck

D1771-25MG
SIALD1771-25MGEA 443 EUR
SIALD1771-25MG
5'-Desoxyadenosin, Sigma-Aldrich®
5'-Desoxyadenosin
Formel: C₁₀H₁₃N₅O₃
CAS-Nummer: 4754-39-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR